Merck
  • Main and secondary relaxations of non-polymeric high-Tg glass formers as revealed by dielectric spectroscopy.
Main and secondary relaxations of non-polymeric high-Tg glass formers as revealed by dielectric spectroscopy.
Physical chemistry chemical physics : PCCP (2020-04-18)
Thomas Körber, Felix Krohn, Christian Neuber, Hans-Werner Schmidt, Ernst A Rössler
ABSTRACT
A series of high-Tg glass formers with Tg values varying between 347 and 390 K and molar masses in the range of 341 and 504 g mol-1 are investigated by dielectric spectroscopy. They are compared to paradigmatic reference systems. Differently polar side groups are attached to a rigid non-polar core unit at different positions. Thereby, the dielectric relaxation strength varies over more than two decades. All the relaxation features typical of molecular glass formers are rediscovered, i.e. stretching of the main (α-) relaxation, a more or less pronounced secondary (β-) process, and a fragility index quite similar to that of other molecular systems. The position of the polar nitrile side group influences the manifestation of the β-relaxation. The α-relaxation stretching displays the trend to become less with higher relaxation strength Δεα, confirming recent reports. Typical for a generic β-process is the increase of its amplitude above Tg, which is found to follow a power-law behaviour as a function of the ratio τα/τβ with a universal exponent; yet, its relative amplitude to that of the α-relaxation varies as does the temporal separation of both processes. The mean activation energy of the β-process as well as the width of the energy distribution gβ(E) increases more or less systematically with Tg. The latter is determined from the dielectric spectra subjected to a scaling procedure assuming a thermally activated process. Plotting gβ(E) as a function of the reduced energy scale E/Tg, the distributions are centred between 19-35 and their widths differ by a factor 2-3.
Sigma-Aldrich
Perhydroisoquinoline, 96%
Sigma-Aldrich
Tripropyl phosphate, 99%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)