Merck
  • Quantification of sphingosine 1-phosphate by validated LC-MS/MS method revealing strong correlation with apolipoprotein M in plasma but not in serum due to platelet activation during blood coagulation.
Quantification of sphingosine 1-phosphate by validated LC-MS/MS method revealing strong correlation with apolipoprotein M in plasma but not in serum due to platelet activation during blood coagulation.
Analytical and bioanalytical chemistry (2015-09-18)
Cecilia Frej, Anders Andersson, Benny Larsson, Li Jun Guo, Eva Norström, Kaisa E Happonen, Björn Dahlbäck
ABSTRACT
Sphingosine 1-phosphate (S1P) is a signalling sphingolipid affecting multiple cellular functions of vascular and immune systems. It circulates at submicromolar levels bound to HDL-associated apolipoprotein M (apoM) or to albumin. S1P in blood is mainly produced by platelets and erythrocytes, making blood sampling for S1P quantification delicate. Standardisation of sampling is thereby of great importance to obtain robust data. By optimising and characterising the extraction procedure and the LC-MS/MS analysis, we have developed and validated a highly specific and sensitive method for S1P quantification. Blood was collected from healthy individuals (n = 15) to evaluate the effects of differential blood sampling on S1P levels. To evaluate correlation between S1P and apoM in different types of plasma and serum, apoM was measured by ELISA. The method showed good accuracy and precision in the range of 0.011 to 0.9 μM with less than 0.07 % carryover. We found that the methanol precipitation used to extract S1P co-extracted apoM and several other HDL-proteins from plasma. The platelet-associated S1P was released during coagulation, thus increasing the S1P concentration to double in serum as compared to that in plasma. Gel filtration chromatography revealed that the platelet-released S1P was mainly bound to albumin. This explains why the strong correlation between S1P and apoM levels in plasma is lost upon the clotting process and hence not observed in serum. We have developed, characterised and validated an efficient, highly sensitive and specific method for the quantification of S1P in biological material.
Product Description
Avanti
Sphingosine-1-Phosphate (d17:1), Avanti Research - A Croda Brand
Pricing and availability is not currently available.