Merck
  • Overexpression of GILZ in macrophages limits systemic inflammation while increasing bacterial clearance in sepsis in mice.
Overexpression of GILZ in macrophages limits systemic inflammation while increasing bacterial clearance in sepsis in mice.
European journal of immunology (2019-12-17)
Mehdi Ellouze, Lola Vigouroux, Colas Tcherakian, Paul-Louis Woerther, Aurélie Guguin, Olivier Robert, Mathieu Surenaud, Thi Tran, Joseph Calmette, Thomas Barbin, Gabriel Perlemuter, Anne-Marie Cassard, Pierre Launay, Virginie Maxime, Djillali Annane, Yves Levy, Véronique Godot
ABSTRACT
Studies support the beneficial effects of glucocorticoids (GCs) during septic shock, steering research toward the potential role of GC-induced proteins in controlling excessive inflammatory responses. GILZ is a glucocorticoid-induced protein involved in the anti-inflammatory effects of GCs. We investigated whether the overexpression of GILZ specifically limited to monocytes and macrophages (M/M) alone could control inflammation, thus improving the outcome of septic shock in animal models. We also monitored the expression of GILZ in M/M from septic mice and septic-shock patients. M/M from patients and septic mice displayed significantly lower expression of GILZ than those isolated from controls. Furthermore, transgenic mice (Tg-mice) experiencing sepsis, with increased expression of GILZ restricted to M/M, showed lower frequencies of inflammatory monocytes than their littermates and lower plasma levels of inflammatory cytokines. Tg-mice also had lower blood bacterial counts. We further established that the upregulation of GILZ in M/M enhanced their phagocytic capacity in in vivo assays. The increase of GILZ in M/M was also sufficient to improve the survival rates of septic mice. These results provide evidence for a central role of both GILZ and M/M in the pathophysiology of septic shock and a possible clue for the modulation of inflammation in this disease.
Sigma-Aldrich
MISSION® shRNA, Lentiviral Particles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Transduction Particles, High Titer, Targets no known mammalian genes