Merck
  • The novel llama-human chimeric antibody has potent effect in lowering LDL-c levels in hPCSK9 transgenic rats.
The novel llama-human chimeric antibody has potent effect in lowering LDL-c levels in hPCSK9 transgenic rats.
Clinical and translational medicine (2020-02-15)
Xinyang Li, Meiniang Wang, Xinhua Zhang, Chuxin Liu, Haitao Xiang, Mi Huang, Yingying Ma, Xiaoyan Gao, Lin Jiang, Xiaopan Liu, Bo Li, Yong Hou, Xiuqing Zhang, Shuang Yang, Naibo Yang
ABSTRACT
The advent of proprotein convertase subtilisin/kexin type 9 (PCSK9)-inhibiting drugs have provided an effective, but extremely expensive treatment for the management of low density lipoprotein (LDL). Our aim was to explore a cost-effective application of camelid anti-PCSK9 single domain antibodies (sdAbs), which are high variable regions of the camelid heavy chain antibodies (VHHs), as a human PCSK9 (hPCSK9) inhibitor. One female llama was immunized with hPCSK9. Screening of high affinity anti-PCSK9 VHHs was carried out based on surface plasmon resonance (SPR) technology. We reported a lysate kinetic analysis method improving the screening efficiency. To increase the serum half-life and targeting properties, the constant region fragment of the human immunoglobulin gamma sub-type 4 (IgG4 Fc) was incorporated to form a novel llama-human chimeric molecule (VHH-hFc). The PCSK9 inhibiting effects of the VHH proteins were analyzed in two human liver hepatocellular cells (HepG2 and Huh7) and in the hPCSK9 transgenic Sprague-Dawley (SD) rat model. The hPCSK9 antagonistic potency of the bivalent VHH-hFc exceeded the monovalent VHH (P < 0.001) in hepatocarcinoma cells. Furthermore, the llama-human chimeric VHH-Fc protein had a similar reduction (~ 40%) of the LDL-c and total cholesterol when compared to the approved evolocumab in transgenic SD rat model, but with low cost. More surprisingly, the chimeric heavy chain antibodies could be persevered for 3 months at room temperature with little loss of the affinity. Due to the high yield and low cost of Pichia pastoris, lipid-lowering effect and strong stability, the llama-human chimeric antibody (VHH-Fc) offers a potent therapeutic candidate for the control of the serum lipid level.
Sigma-Aldrich
Freund′s Adjuvant, Incomplete, liquid
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Millipore
PES Membrane Filter, 0.22 μm Pore Size, Millipore Express®, filter diam. 25 mm, hydrophilic