Merck
  • TNFalpha-induced GM-CSF release from human airway smooth muscle cells depends on activation of an ET-1 autoregulatory positive feedback mechanism.
TNFalpha-induced GM-CSF release from human airway smooth muscle cells depends on activation of an ET-1 autoregulatory positive feedback mechanism.
Thorax (2009-10-24)
J Knobloch, H Peters, D Jungck, K Müller, J Strauch, A Koch
ABSTRACT
There is an urgent need to inhibit endothelin-1 (ET-1) induced chronic inflammatory processes in early stages of lung diseases in order to prevent untreatable irreversible stages often accompanied by lung fibrosis and pulmonary hypertension. Nothing is known about the airway inflammation-inducing and/or maintaining role of ET-1 in human airway smooth muscle cells (HASMCs). ET-1 and granulocyte-macrophage colony-stimulating factor (GM-CSF) expression in response to tumour necrosis factor alpha (TNFalpha) and ET-1 stimulation was investigated, and the impact of mitogen-activated protein kinase (MAPK) pathways in this context was studied. To elucidate the anti-inflammatory properties of the dual endothelin receptor antagonist bosentan that targets both endothelin receptor subtypes A (ET(A)R) and B (ET(B)R), its effect on the TNFalpha/ET-1/GM-CSF network was investigated. ET-1 and GM-CSF expression and activation of MAPKs were investigated via quantitative reverse transcription-PCR (RT-PCR), western blotting and ELISA. Both TNFalpha and ET-1 activated p38(MAPK) and extracellular signal-regulated kinase (ERK)-1/-2 signalling. ET-1 expression was induced by TNFalpha and by ET-1 itself. Both effects were inhibited by bosentan and by specific ET(A)R or p38(MAPK) blockade. ET-1- and TNFalpha-induced GM-CSF expression were both reduced by bosentan as well as by specific inhibition of either ET(A)R, ET(B)R, p38(MAPK) or ERK-1/-2. TNFalpha activates an ET(A)R- and p38(MAPK)-dependent ET-1 autoregulatory positive feedback loop to maintain GM-CSF release from HASMCs. Since bosentan impairs ET-1 autoregulation and TNFalpha-induced ET-1 release, as well as TNFalpha- and ET-1-induced GM-CSF release, the present data suggest therapeutic utility for bosentan in treating particularly the early stages of chronic inflammatory airway diseases.
Sigma-Aldrich
BQ-123, ≥99%, sodium salt, lyophilized powder
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.