Merck
  • Integrin-generated forces lead to streptavidin-biotin unbinding in cellular adhesions.
Integrin-generated forces lead to streptavidin-biotin unbinding in cellular adhesions.
Biophysical journal (2014-04-08)
Carol Jurchenko, Yuan Chang, Yoshie Narui, Yun Zhang, Khalid S Salaita
ABSTRACT
The interplay between chemical and mechanical signals plays an important role in cell biology, and integrin receptors are the primary molecules involved in sensing and transducing external mechanical cues. We used integrin-specific probes in molecular tension fluorescence microscopy to investigate the pN forces exerted by integrin receptors in living cells. The molecular tension fluorescence microscopy probe consisted of a cyclic Arg-Gly-Asp-D-Phe-Lys(Cys) (cRGDfK(C)) peptide tethered to the terminus of a polyethylene glycol polymer that was attached to a surface through streptavidin-biotin linkage. A fluorescence resonance energy transfer mechanism was used to visualize tension-driven extension of the polymer. Surprisingly, we found that integrin receptors dissociate streptavidin-biotin tethered ligands in focal adhesions within 60 min of cell seeding. Although streptavidin-biotin binding affinity is described as the strongest noncovalent bond in nature, and is ~10(6) - 10(8) times larger than that of integrin-RGD affinity, our results suggest that individual integrin-ligand complexes undergo a marked enhancement in stability when the receptor assembles in the cell membrane. Based on the observation of streptavidin-biotin unbinding, we also conclude that the magnitude of integrin-ligand tension in focal adhesions can reach values that are at least 10 fold larger than was previously estimated using traction force microscopy-based methods.
Sigma-Aldrich
Anti-Vinculin−FITC antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Pricing and availability is not currently available.