Merck
  • Differential regulation of the attachment of Kaposi's sarcoma-associated herpesvirus (KSHV)-infected human B cells to extracellular matrix by KSHV-encoded gB and cellular alphaV integrins.
Differential regulation of the attachment of Kaposi's sarcoma-associated herpesvirus (KSHV)-infected human B cells to extracellular matrix by KSHV-encoded gB and cellular alphaV integrins.
Cellular microbiology (2008-03-28)
Ossie F Dyson, Telisha L Oxendine, Khalief E Hamden, Patrick W Ford, Shaw M Akula
ABSTRACT
Kaposi's sarcoma-associated herpesvirus (KSHV) has two modes of replications: latent and lytic replications. Reactivation from latency is dictated, in part, by the cell cycle. Herein, we have attempted to delineate the importance of cell cycle in KSHV pathogenesis by exploring the expression pattern of cell-surface receptors during different phases of the cell cycle. alphaV integrin expression is augmented during S phase in fibroblasts, epithelial and KSHV-infected cells. Using a Matrigel system, we pioneer the concept that KSHV-infected primary effusion lymphoma cells can attach to extracellular matrix proteins. This attachment is mediated primarily via alphaV integrins or virally encoded gB, and occurs preferentially in cells from S phase or cells from S phase actively supporting a lytic infection respectively. Such an ability of infected B cells to attach to endothelial cells may also aid in the dissemination of infection. The keystone of this work is that for the first time, we describe the ability of KSHV-infected B cells to preferentially use cellular (alphaV) or viral (gB) receptors to specifically bind cells, depending upon the stage of the cell cycle and infection.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, clone 6S6, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid