Merck
  • Polarization of Human Macrophages by Interleukin-4 Does Not Require ATP-Citrate Lyase.
Polarization of Human Macrophages by Interleukin-4 Does Not Require ATP-Citrate Lyase.
Frontiers in immunology (2018-12-21)
Dmitry Namgaladze, Sven Zukunft, Frank Schnütgen, Nina Kurrle, Ingrid Fleming, Dominik Fuhrmann, Bernhard Brüne
ABSTRACT
Macrophages exposed to the Th2 cytokines interleukin (IL) IL-4 and IL-13 exhibit a distinct transcriptional response, commonly referred to as M2 polarization. Recently, IL-4-induced polarization of murine bone marrow-derived macrophages (BMDMs) has been linked to acetyl-CoA levels through the activity of the cytosolic acetyl-CoA-generating enzyme ATP-citrate lyase (ACLY). Here, we studied how ACLY regulated IL-4-stimulated gene expression in human monocyte-derived macrophages (MDMs). Although multiple ACLY inhibitors attenuated IL-4-induced target gene expression, this effect could not be recapitulated by silencing ACLY expression. Furthermore, ACLY inhibition failed to alter cellular acetyl-CoA levels and histone acetylation. We generated ACLY knockout human THP-1 macrophages using CRISPR/Cas9 technology. While these cells exhibited reduced histone acetylation levels, IL-4-induced gene expression remained intact. Strikingly, ACLY inhibitors still suppressed induction of target genes by IL-4 in ACLY knockout cells, suggesting off-target effects of these drugs. Our findings suggest that ACLY may not be the major regulator of nucleocytoplasmic acetyl-CoA and IL-4-induced polarization in human macrophages. Furthermore, caution should be warranted in interpreting the impact of pharmacological inhibition of ACLY on gene expression.
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, serum, Upstate®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium butyrate, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
SB-204990, ≥98% (HPLC)