Merck
  • Inhibition of GSK3β improves hippocampus-dependent learning and rescues neurogenesis in a mouse model of fragile X syndrome.
Inhibition of GSK3β improves hippocampus-dependent learning and rescues neurogenesis in a mouse model of fragile X syndrome.
Human molecular genetics (2011-11-04)
Weixiang Guo, Adeline C Murthy, Li Zhang, Eric B Johnson, Eric G Schaller, Andrea M Allan, Xinyu Zhao
ABSTRACT
Fragile X syndrome (FXS), a common inherited form of intellectual disability with learning deficits, results from a loss of fragile X mental retardation protein (FMRP). Despite extensive research, treatment options for FXS remain limited. Since FMRP is known to play an important role in adult hippocampal neurogenesis and hippocampus-dependent learning and FMRP regulates the adult neural stem cell fate through the translational regulation of glycogen synthase kinase 3β (GSK3β), we investigated the effects of a GSK3β inhibitor, SB216763, on Fmr1 knockout mice (Fmr1 KO). We found that the inhibition of GSK3β could reverse the hippocampus-dependent learning deficits and rescue adult hippocampal neurogenesis at multiple stages in Fmr1 KO mice. Our results point to GSK3β inhibition as a potential treatment for the learning deficits seen in FXS.
Sigma-Aldrich
Anti-c-Fos Antibody, Chemicon®, from sheep
Sigma-Aldrich
Anti-Fragile X Mental Retardation Protein Antibody, clone 1C3, ascites fluid, clone 1C3, Chemicon®
Sigma-Aldrich
Anti-β-Catenin Antibody, clone 2H4A7, clone 2H4A7, Upstate®, from mouse
Sigma-Aldrich
SB 216763, >98% (HPLC)