Merck
  • Characterization of polysaccharide production of haemophilus influenzae Type b and its relationship to bacterial cell growth.
Characterization of polysaccharide production of haemophilus influenzae Type b and its relationship to bacterial cell growth.
Applied biochemistry and biotechnology (2003-09-30)
Mickie Takagi, Joaquin Cabrera-Crespo, Júlia Baruque-Ramos, Teresa Cristina Zangirolami, Isaias Raw, Martha Massako Tanizaki
ABSTRACT
Haemophilus influenzae type b (Hib) causes invasive infections in infants and young children. Vaccines consisting of Hib capsular polysaccharide (polymer of ribosylribitol phosphate [PRP]) conjugated to a protein are effective in the prevention of such infections. The production of capsular polysaccharide type b was studied in three cultivation conditions: single, glucose pulse, and repeated batch. Specific polysaccharide production (Yp/x) was calculated for all experiments, showing the following values: 67 (single-batch cultivation), 71 (glucose pulse), 75 (repeated-batch cultivation, first batch), and 87 mg of PRP/g of dry cell weight (DCW) (repeated-batch cultivation, second batch). Biomass concentration reached approximately 1.8 g of DCW/L, while polysaccharide concentration was about approximately 132 mg/L in the three fermentation runs. Polysaccharide synthesis is associated with cell growth in all studied conditions as established by Kono's analysis and Luedeking-Piret's model.
Sigma-Aldrich
RIPAb+ Ago2 - RIP Validated Antibody and Primer Set, ascites fluid, from mouse