Merck
  • Cloning of mouse immunoglobulin epsilon gene and its location within the heavy chain gene cluster.
Cloning of mouse immunoglobulin epsilon gene and its location within the heavy chain gene cluster.
Proceedings of the National Academy of Sciences of the United States of America (1981-03-01)
Y Nishida, T Kataoka, N Ishida, S Nakai, T Kishimoto, I Böttcher, T Honjo
ABSTRACT
Mouse immunoglobulin epsilon chain gene was cloned from DNA of a hybridoma producing anti-dinitrophenyl IgE, which was constructed by fusing a spleen cell of a BALB/c mouse with a variant clone of MOPC21 myeloma (IgG1 producer). Because a given active heavy chain constant region (CH) gene is linked to a heavy chain joining segment (JH) gene at its 5' side, the expressed C epsilon gene of the hybridoma was cloned from a phage library containing partial Sau3A digests of IgE hybridoma DNA by using a J gene fragment as a probe. Among 6 X 10(5) phages screened, five positive clones were obtained and three of them were identified as C epsilon gene clones by restriction mapping, Southern blot hybridization, R-loop formation, and partial nucleotide sequence determination. The determined nucleotide sequence predicted the amino acid sequence which resembles a part of the CH3 domain of human epsilon chain. The deletion profile of the C epsilon gene in various myelomas expressing different CH genes indicates that the C epsilon gene is located between the C gamma 2a and C alpha genes. The linkage (5'-epsilon-alpha-3') was directly confirmed by molecular cloning of the overlapping chromosomal segments from newborn mouse DNA.
Sigma-Aldrich
Citrate Synthase from porcine heart, ammonium sulfate suspension, ≥100 units/mg protein