Merck
  • Regioselective Synthesis of Fluorosulfonyl 1,2,3-Triazoles from Bromovinylsulfonyl Fluoride.
Regioselective Synthesis of Fluorosulfonyl 1,2,3-Triazoles from Bromovinylsulfonyl Fluoride.
Organic letters (2018-06-16)
Joice Thomas, Valery V Fokin
ABSTRACT
A regioselective metal-free preparation of 4-fluorosulfonyl 1,2,3-triazoles from organic azides and a hitherto underexplored bromovinylsulfonyl fluoride building block is described. This reaction is very general and was extended to the synthesis of various sulfonates, sulfonamides, and sulfonic acid derivatives of triazoles and other azole heterocycles which would otherwise be difficult to access by existing methods.
Sigma-Aldrich
1,2-Dibromoethane-1-sulfonyl fluoride
Pricing and availability is not currently available.