Skip to Content
Fechas de fin de año: Facturación de pedidos hasta 13/12. Receso de Merck Argentina del 23/12 al 02/01.
Merck
  • In vivo screening identifies GATAD2B as a metastasis driver in KRAS-driven lung cancer.
In vivo screening identifies GATAD2B as a metastasis driver in KRAS-driven lung cancer.
Nature communications (2018-07-18)
Caitlin L Grzeskowiak, Samrat T Kundu, Xiulei Mo, Andrei A Ivanov, Oksana Zagorodna, Hengyu Lu, Richard H Chapple, Yiu Huen Tsang, Daniela Moreno, Maribel Mosqueda, Karina Eterovic, Jared J Fradette, Sumreen Ahmad, Fengju Chen, Zechen Chong, Ken Chen, Chad J Creighton, Haian Fu, Gordon B Mills, Don L Gibbons, Kenneth L Scott
ABSTRACT
Genetic aberrations driving pro-oncogenic and pro-metastatic activity remain an elusive target in the quest of precision oncology. To identify such drivers, we use an animal model of KRAS-mutant lung adenocarcinoma to perform an in vivo functional screen of 217 genetic aberrations selected from lung cancer genomics datasets. We identify 28 genes whose expression promoted tumor metastasis to the lung in mice. We employ two tools for examining the KRAS-dependence of genes identified from our screen: 1) a human lung cell model containing a regulatable mutant KRAS allele and 2) a lentiviral system permitting co-expression of DNA-barcoded cDNAs with Cre recombinase to activate a mutant KRAS allele in the lungs of mice. Mechanistic evaluation of one gene, GATAD2B, illuminates its role as a dual activity gene, promoting both pro-tumorigenic and pro-metastatic activities in KRAS-mutant lung cancer through interaction with c-MYC and hyperactivation of the c-MYC pathway.
Sigma-Aldrich
Eosin Y Solution, Alcoholic
Pricing and availability is not currently available.