Merck
  • Amyloid Beta and Tau as Alzheimer's Disease Blood Biomarkers: Promise From New Technologies.
Amyloid Beta and Tau as Alzheimer's Disease Blood Biomarkers: Promise From New Technologies.
Neurology and therapy (2017-07-25)
Lih-Fen Lue, Andre Guerra, Douglas G Walker
ABSTRACT
The utility of the levels of amyloid beta (Aβ) peptide and tau in blood for diagnosis, drug development, and assessment of clinical trials for Alzheimer's disease (AD) has not been established. The lack of availability of ultra-sensitive assays is one critical issue that has impeded progress. The levels of Aβ species and tau in plasma and serum are much lower than levels in cerebrospinal fluid. Furthermore, plasma or serum contain high levels of assay-interfering factors, resulting in difficulties in the commonly used singulex or multiplex ELISA platforms. In this review, we focus on two modern immune-complex-based technologies that show promise to advance this field. These innovative technologies are immunomagnetic reduction technology and single molecule array technology. We describe the technologies and discuss the published studies using these technologies. Currently, the potential of utilizing these technologies to advance Aβ and tau as blood-based biomarkers for AD requires further validation using already collected large sets of samples, as well as new cohorts and population-based longitudinal studies.
Sigma-Aldrich
Anti-β-Amyloid antibody, Mouse monoclonal, clone BAM-10, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Tau antibody, Mouse monoclonal, clone Tau46, purified from hybridoma cell culture
Pricing and availability is not currently available.