Zinc isopropylsulfinate
Synonym(s):
Baran IPS Reagent, Bis((isopropylsulfinyl)oxy)zinc, IPS
Empirical Formula (Hill Notation):
C6H14O4S2Zn
CAS Number:
Molecular Weight:
279.69
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing