
Vinylene carbonate

Synonym(s):
1,3-Dioxol-2-one, VC
Empirical Formula (Hill Notation):
C3H2O3
CAS Number:
Molecular Weight:
86.05
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing