
Valrubicin

Synonym(s):
Trifluoroacetyladriamycin 14-valerate
Empirical Formula (Hill Notation):
C34H36F3NO13
CAS Number:
Molecular Weight:
723.64
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing