
Tyrphostin 1

Synonym(s):
(4-Methoxybenzylidene)malononitrile
Linear Formula:
CH3OC6H4CH=C(CN)2
CAS Number:
Molecular Weight:
184.19
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing