Skip to Content
Merck
Trimethyl phosphate ≥99%

Trimethyl phosphate

Trimethyl phosphate ≥99%
Synonym(s):
TMP, TMPA, TMPO, Trimethyl phosphate
Linear Formula:
(CH3)3PO4
CAS Number:
Molecular Weight:
140.07

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing