Tributyl citrate
Synonym(s):
Tributyl citrate
Linear Formula:
HOC(COOCH2CH2CH2CH3)(CH2COOCH2CH2CH2CH3)2
CAS Number:
Molecular Weight:
360.44
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing