
Thymol solution

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing