Skip to Content
Merck
Tetrahydrofurfuryl alcohol 99%

Tetrahydrofurfuryl alcohol

Tetrahydrofurfuryl alcohol 99%
Synonym(s):
NSC 15434, Tetrahydro-2-furanmethanol, Tetrahydrofurfuryl alcohol
Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing