Tetrahydro-2H-thiopyran-4-ol 1,1-dioxide
Empirical Formula (Hill Notation):
C5H10O3S
CAS Number:
Molecular Weight:
150.20
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing