Skip to Content
Merck
Tetrabutylammonium tribromide 98%

Tetrabutylammonium tribromide

Tetrabutylammonium tribromide 98%
Synonym(s):
TBABr3, Tetrabutylammonium bromide perbromide
Linear Formula:
(CH3CH2CH2CH2)4NBr3
CAS Number:
Molecular Weight:
482.18

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing