Skip to Content
Merck
tert-Butyl propiolate 98%

tert-Butyl propiolate

tert-Butyl propiolate 98%
Synonym(s):
tert-Butyl acetylenecarboxylate
Linear Formula:
HC≡CCOOC(CH3)3
CAS Number:
Molecular Weight:
126.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing