
Suxamethonium chloride

Synonym(s):
Succinylcholine chloride dihydrate, Bis(trimethylammonioethyl) succinate chloride, Succinyldicholine dichloride
Linear Formula:
[(CH3)3NCH2CH2OCOCH2]2Cl2 · 2H2O
CAS Number:
Molecular Weight:
397.34
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing