
Sulfobutylbetadex impurity B

Synonym(s):
1,4-Butane sultone, 4-Hydroxybutane-1-sulfonic acid δ-sultone
Empirical Formula (Hill Notation):
C4H8O3S
CAS Number:
Molecular Weight:
136.17
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing