Skip to Content
Merck
Spiroxamine PESTANAL®, analytical standard

Spiroxamine

Spiroxamine PESTANAL®, analytical standard
Synonym(s):
N-Ethyl-N-propyl-8-tert-butyl-1,4-dioxaspiro[4.5]dec-2-ylmethylamine
Empirical Formula (Hill Notation):
C18H35NO2
CAS Number:
Molecular Weight:
297.48

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique