
Sodium triacetoxyborohydride

Synonym(s):
STAB, Sodium triacetoxyborohydride
Linear Formula:
(CH3COO)3BHNa
CAS Number:
Molecular Weight:
211.94
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing