
Sodium Phosphate, Monobasic

Synonym(s):
Sodium phosphate monobasic monohydrate, Monosodium phosphate, Sodium dihydrogen phosphate monohydrate
Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
Products
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing