
Sodium pentanesulfonate

Synonym(s):
1-Pentanesulfonic acid sodium salt
Linear Formula:
C5H11O3SNa
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing