
Silica, fumed

Synonym(s):
Silica, Silicic anhydride, Silicon dioxide, Silicon dioxide amorphous
Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
Products
Brand
Formula Weight
Physical Form
Quality Segment
Particle Size (µm)
Showing 1-2 of 2
Compare
Product Number
Description
Pricing