![RPMI-1640 Medium With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/214/387/07dca72b-593a-422f-a631-b84346835e6a/640/07dca72b-593a-422f-a631-b84346835e6a.jpg)
RPMI-1640 Medium
![RPMI-1640 Medium With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/214/387/07dca72b-593a-422f-a631-b84346835e6a/640/07dca72b-593a-422f-a631-b84346835e6a.jpg)
Synonym(s):
RPMI-1640 Medium
Products
Product Category
Brand
Physical Form
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Sodium Pyruvate (g/L)
Stable Glutamine
Quality Segment
Sterility
Sterilization Method
Technique
Showing 1-20 of 21
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2