
Rivastigmine

Synonym(s):
N-Ethyl-N-methylcarbamic acid 3-[(1S)-1-(dimethylamino)ethyl]phenyl-ester
Empirical Formula (Hill Notation):
C14H22N2O2
CAS Number:
Molecular Weight:
250.34
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing