
Retinol solution

Synonym(s):
Retinol solution, 3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexenyl)-2,4,6,8-nonatetraen-1-ol, Axerophthol, Vitamin A alcohol solution, Vitamin A1, all-trans-Retinol
Empirical Formula (Hill Notation):
C20H30O
CAS Number:
Molecular Weight:
286.45
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing