
(R)(−)-α-Methylhistamine dihydrochloride

Synonym(s):
(R)(−)-α-Methyl-1H-imidazole-4-ethanamine dihydrochloride
Empirical Formula (Hill Notation):
C6H11N3 · 2HCl
CAS Number:
Molecular Weight:
198.09
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing