![Propyl gallate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/263/500/e9fa4b9c-b3bf-480a-9a4f-25493518d58c/640/e9fa4b9c-b3bf-480a-9a4f-25493518d58c.png)
Propyl gallate
![Propyl gallate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/263/500/e9fa4b9c-b3bf-480a-9a4f-25493518d58c/640/e9fa4b9c-b3bf-480a-9a4f-25493518d58c.png)
Synonym(s):
3,4,5-Trihydroxybenzoic acid propyl ester
Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing