
Propisochlor

Synonym(s):
2-Chloro-2′-ethyl-6′-methyl-N-(isopropoxymethyl)acetanilide, Proponit
Empirical Formula (Hill Notation):
C15H22ClNO2
CAS Number:
Molecular Weight:
283.79
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing