Skip to Content
Merck
Potassium benzoate ReagentPlus®, 99%

Potassium benzoate

Potassium benzoate ReagentPlus®, 99%
Synonym(s):
Benzoic acid potassium salt
Linear Formula:
C6H5COOK
CAS Number:
Molecular Weight:
160.21
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3