
Platinum(II) 2,3,7,8,12,13,17,18-octaethyl-21H,23H-porphyrin

Synonym(s):
Platinum octaethylporphyrin, Platinum(II) 2,3,7,8,12,13,17,18-octaethyl-21H,23H-porphyrin, PtOEP
Empirical Formula (Hill Notation):
C36H44N4Pt
CAS Number:
Molecular Weight:
727.84
Products
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing