Skip to Content
Merck
Phosphorus(V) oxychloride 99.999%

Phosphorus(V) oxychloride

Phosphorus(V) oxychloride 99.999%
Synonym(s):
Phosphorus(V) oxide chloride, Phosphoryl chloride
Linear Formula:
POCl3
CAS Number:
Molecular Weight:
153.33
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing