Perfluorodecanoic acid
Synonym(s):
Nonadecafluorodecanoic acid, Nonadecafluorocapric acid, Perfluorocapric acid, Perfluorodecanoic acid
Linear Formula:
CF3(CF2)8CO2H
CAS Number:
Molecular Weight:
514.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Agency
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3