
Ondansetron Related Compound A

Synonym(s):
3-[(Dimethylamino)methyl]-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one hydrochloride, 3-[(Dimethylamino)methyl]-1,2,3,9-tetrahydro-9-methyl-4H-carbazol-4-one
Empirical Formula (Hill Notation):
C16H20N2O · HCl
CAS Number:
Molecular Weight:
292.80
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2