
Ondansetron for LC system suitability

Synonym(s):
Ondansetron hydrochloride dihydrate, 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one hydrochloride, GR 38032F
Empirical Formula (Hill Notation):
C18H19N3O · HCl · 2H2O
CAS Number:
Molecular Weight:
365.85
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing