
Octyl β-D-1-thioglucopyranoside

Synonym(s):
Octyl thioglucoside, n-Octyl-β-D-thioglucopyranoside, ULTROL® Grade
Empirical Formula (Hill Notation):
C14H28O5S
CAS Number:
Molecular Weight:
308.43
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2