
Nutrient Broth

Synonym(s):
Standard - Nutrient Broth
Products
Product Category
Brand
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing