![Norethisterone for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/159/388/ec309ce5-6f7f-4e4d-829a-665d3b7071fc/640/ec309ce5-6f7f-4e4d-829a-665d3b7071fc.png)
Norethisterone for system suitability
![Norethisterone for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/159/388/ec309ce5-6f7f-4e4d-829a-665d3b7071fc/640/ec309ce5-6f7f-4e4d-829a-665d3b7071fc.png)
Synonym(s):
19-Norethindrone, 17α-Ethynyl-19-nortestosterone, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one, 17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one, 19-Nor-17α-ethynyl-4-androsten-17β-ol-3-one, 19-Norethisterone
Empirical Formula (Hill Notation):
C20H26O2
CAS Number:
Molecular Weight:
298.42
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing