Skip to Content
Merck
N,N-Dimethylmethyleneiminium chloride technical grade, 90%

N,N-Dimethylmethyleneiminium chloride

N,N-Dimethylmethyleneiminium chloride technical grade, 90%
Synonym(s):
Dimethylformiminium chloride, Methylenedimethylammonium chloride
Linear Formula:
CH2=N+(CH3)2Cl-
CAS Number:
Molecular Weight:
93.56
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Reaction Type
Special Grade
Showing 1-2 of 2