N-Maleoyl-β-alanine
Synonym(s):
3-Maleimidopropionic acid, N-(2-Carboxyethyl)maleimide
Empirical Formula (Hill Notation):
C7H7NO4
CAS Number:
Molecular Weight:
169.13
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing