
N-Heptafluorobutyrylimidazole

Synonym(s):
1-(Perfluorobutyryl)imidazole, NSC 151966
Empirical Formula (Hill Notation):
C7H3F7N2O
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing